Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB40145A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB40145A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB40145A750 Supplier Novus Biologicals Supplier No. NB400145AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD36 Polyclonal specifically detects CD36 in Human, Mouse, Amphibian samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoblotting.

Specifications

Antigen CD36/SR-B3
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor
Gene Symbols CD36
Host Species Rabbit
Immunogen A synthetic peptide mapping to a region of human CD36 between residues 300-400. [UniProtNo. P16671]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 948
Target Species Human, Mouse, Amphibian
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.