Learn More
Description
Specifications
Specifications
| Antigen | CD36/SR-B3 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000-1:10000, Simple Western 1:40, Flow Cytometry reported in scientific literature (PMID 32971872), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27226602), Immunoblotting reported in scientific literature (PMID 31501473), SDS-Page reported in scientific literature (PMID 31501473) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | P16671 |
| Gene Alias | CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor |
| Gene Symbols | CD36 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
