Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD36 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
$206.50 - $501.50
Specifications
Antigen | CD36/SR-B3 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:2000-1:10000, Simple Western 1:40, Flow Cytometry reported in scientific literature (PMID 32971872), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27226602), Immunoblotting reported in scientific literature (PMID 31501473), SDS-Page reported in scientific literature (PMID 31501473) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400145SS
![]() |
Novus Biologicals
NB400145SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB400145
![]() |
Novus Biologicals
NB400145 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Amphibian samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting, SDS-Page.Specifications
CD36/SR-B3 | |
Western Blot 1:2000-1:10000, Simple Western 1:40, Flow Cytometry reported in scientific literature (PMID 32971872), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27226602), Immunoblotting reported in scientific literature (PMID 31501473), SDS-Page reported in scientific literature (PMID 31501473) | |
Polyclonal | |
Rabbit | |
Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
CD36 | |
IgG | |
Affinity Purified | |
Human.Mouse reactivity reported in scientific literature (PMID: 24163073) Immunogen sequence has 95% homology to rat, monkey, Golden hamster and rabbit, 90% homology to mouse, and 75% homology to bovine. |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot | |
Unconjugated | |
RUO | |
Human, Mouse, Amphibian | |
P16671 | |
948 | |
A synthetic peptide mapping to a region of human CD36 between residues 300-400. [UniProt# P16671] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
75 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title