Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36 Antibody (D-2712) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11059724T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11059724T 0.025 mL
NB11059724 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11059724T Supplier Novus Biologicals Supplier No. NB11059724SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 8 publications

CD36 Monoclonal specifically detects CD36 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen CD36/SR-B3
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone D-2712
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:100-1:250, Immunoprecipitation 10-20 ug, Immunohistochemistry-Paraffin 1:100-1:250
Gene Alias CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor
Gene Symbols CD36
Host Species Mouse
Immunogen Adenovirus expressing recombinant mouse CD36. [Uniprot: Q08857]
Molecular Weight of Antigen 75 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 948
Test Specificity Mouse and rat. Does not react with human.
Target Species Mouse, Rat, Human (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgA κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.