Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36/SR-B3 Antibody (255606), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB175460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175460 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175460 Supplier Novus Biologicals Supplier No. FAB19551Z
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD36/SR-B3 Monoclonal antibody specifically detects CD36/SR-B3 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD36/SR-B3
Applications Flow Cytometry
Classification Monoclonal
Clone 255606
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, Thrombospondin receptor
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD36/SR-B3, Gly30-Asn439, Accession # P16671
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 948
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.