Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36/SR-B3 Antibody (324205), PerCP, Novus Biologicals™
SDP

Catalog No. NB120426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120426 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120426 Supplier Novus Biologicals Supplier No. FAB25191C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD36/SR-B3 Monoclonal specifically detects CD36/SR-B3 in Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CD36/SR-B3
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 324205
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, Thrombospondin receptor
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD36/SR-B3, Gly30-Lys439, Accession # Q08857
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 948
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.