Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD37 Antibody (424925R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB369573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369573 Supplier Novus Biologicals Supplier No. FAB4625RI
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD37 Monoclonal specifically detects CD37 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CD37
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 424925R
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD37, Accession # Q96LM7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 951
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.