Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD37 Antibody (IPO-24), Novus Biologicals™

Mouse Monoclonal Antibody
$246.61 - $542.38
Specifications
| Antigen | CD37 |
|---|---|
| Clone | IPO-24 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, SDS-Page, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24475000
![]() |
Novus Biologicals
NBP2447500.02MG |
0.02 mg |
Each for $246.61
|
|
|||||
NBP24475001
![]() |
Novus Biologicals
NBP2447500.1MG |
0.1 mg |
Each for $542.38
|
|
|||||
NBP24475002
![]() |
Novus Biologicals
NBP2447500.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD37 Monoclonal specifically detects CD37 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, Immunofluorescence.Specifications
| CD37 | |
| 0.2 mg/mL | |
| Flow Cytometry, SDS-Page, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Immunology | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 951 | |
| Spleen cells of a patient with hairy cell leukemia | |
| Primary | |
| Store at 4C. |
| IPO-24 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
| CD37 | |
| IgG2b κ | |
| Protein A or G purified | |
| Recognizes a protein of 33-55kDa, identified as CD37 (Workshop V; Code CD37.7). CD37 is strongly expressed on normal and neoplastic mature (sIg+) B-lymphocytes. In B-cell ontogeny, CD37 appears after the pre-B-cell stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells.1 CD37 is also present, at low densities, on resting and activated T cells, neutrophils, monocytes, and some myelomonocytic leukemia cells. It is absent from platelets, erythrocytes. CD37 is a member of a family of tetraspan transmembrane proteins, including CD9, CD53, CD63, CD81, and CD82. It associates other tetraspan transmembrane proteins and MHC class II molecules to form a large complex at the surface of B cells and play a role in signal transduction. CD37 is a valuable and stable marker for peripheral mature B-cells and corresponding malignancies like B-cell chronic lymphocytic leukemia (B-CLL), hairy cell leukemia (HCL), and all types of B-cell non-Hodgkin's lymphoma (B-NHL). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title