Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Antibody (1G7F4), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP225265Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225265Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225265Z Supplier Novus Biologicals Supplier No. NBP225265AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD38 Monoclonal specifically detects CD38 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD38
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1G7F4
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Gene Symbols CD38
Host Species Mouse
Immunogen Amino acids 71-197 of mouse CD38 were used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 952
Test Specificity The amino acid sequence used as immunogen for the antibody is 100% homologous in Mouse, 91% in Rat and 64% in Human.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.