Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Antibody (HIT2), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N50510PECY5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N50510PECY5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N50510PECY5 Supplier Novus Biologicals Supplier No. NB500510PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD38
Applications Flow Cytometry
Classification Monoclonal
Clone HIT2
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Gene Symbols CD38
Host Species Mouse
Immunogen Human thymocytes in foetus
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 952.0
Test Specificity The antibody HIT2 reacts with CD38 (T10), a 45 kDa type II transmembrane glycoprotein strongly expressed mainly on plasma cells and activated T and B lymphocytes; it is an antigenic marker of lymphoid cells.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.