Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Mouse anti-Human, BV650, Clone: HB7 (also known as HB-7), BD Horizon™

Mouse Monoclonal Antibody

$225.00 - $380.00

Specifications

Antigen CD38
Clone HB7 (also known as HB-7)
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Violet 650
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB569385
View Documents
BD Biosciences
569385
25 Tests
Each for $225.00
Only null left
Add to Cart
 
BDB569966
View Documents
BD Biosciences
569966
100 Tests
Each for $380.00
Only null left
Add to Cart
 
Description

Description

CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow cytometry
Specifications

Specifications

CD38
Flow Cytometry
Brilliant Violet 650
Mouse
Human
T10; ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; OKT10
Human BJAB B cell line
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
HB7 (also known as HB-7)
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
952
IgG1 κ
Affinity Chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.