Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD39/ENTPD1 Antibody (004), Alexa Fluor™ 532
SDP

Catalog No. NB075056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075056 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075056 Supplier Novus Biologicals Supplier No. NBP290564AF532

Rabbit Monoclonal Antibody

CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Mouse samples. It is validated for ELISA.

Specifications

Antigen CD39/ENTPD1
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD39/ENTPD1 (P55772; Thr38-Ile478).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 953
Test Specificity No cross-reactivity in ELISA with: Human ENTPD3 Human ENTPD5
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.