Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD39/ENTPD1 Antibody (495826), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB370170 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370170 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370170 Supplier Novus Biologicals Supplier No. FAB4398D
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen CD39/ENTPD1
Applications CyTOF, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 495826
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD39/ENTPD1, Thr38-Ile478, Accession # AAH11278
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 953
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.