Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD39L4/ENTPD5 Rabbit anti-Human, Clone: 64, Novus Biologicals™
SDP

Catalog No. NB021129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021129 100 μg
NB021130 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021129 Supplier Novus Biologicals Supplier No. NBP289830100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD39L4/ENTPD5 Monoclonal specifically detects CD39L4/ENTPD5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Sandwich ELISA Capture.

Specifications

Antigen CD39L4/ENTPD5
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone 64
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry, Immunoprecipitation 0.2-1 ul/mg of lysate, Immunohistochemistry-Paraffin 1:500-1:2000, Sandwich ELISA Capture 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias CD39 antigen-like 4, CD39L4NTPDase 5, CD39-like 4, EC 3.6.1, EC 3.6.1.42, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 5, ER-UDPase, GDPase ENTPD5, Guanosine-diphosphatase ENTPD5, MGC163357, MGC163359, NTPDase-5, Nucleoside diphosphatase, PCPH, Pcph proto-oncogene protein, proto-oncogene CPH, UDPase ENTPD5, Uridine-diphosphatase ENTPD5
Host Species Rabbit
Immunogen Recombinant Human CD39L4/ENTPD5 protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 957
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.