Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD4 Ab-8, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry procedures with Epredia™ CD4 Ab-8, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS1528S

 View more versions of this product

Catalog No. MS1528S


Only null left
Add to Cart

Description

Description

CD4, a single chain transmembrane glycoprotein, is found on a T cell subset (helper/inducer) representing 45% of peripheral blood lymphocytes. It is also present on 80% of thymocytes and at a lower level on monocytes. It is involved in recognition of antigen presented along with MHC class II by APCs. It serves as receptor for HIV.

Host Species: Mouse

Clone: 4B12

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 59 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD4 Ab-8
Monoclonal
Unconjugated
Mouse
1 mL
T-Cell
Human
Concentrated
Immunohistochemistry (Paraffin)
4B12
1:10-1:20 for 60 min at RT
Stimulated human leukocytes
IVD
Primary
Store vial at 4deg.C. When stored at 2 to 8deg.C,
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.