Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD4 Ab-8, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry procedures with Epredia™ CD4 Ab-8, Mouse Monoclonal Antibody.

$449.00 - $1866.00

Specifications

Antigen CD4 Ab-8
Clone 4B12
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS1528S0
View Documents
Epredia™
MS1528S0
Concentrated 100 μL
Each for $449.00
Only null left
Add to Cart
 
MS1528S1
View Documents
Epredia™
MS1528S1
Concentrated 500 μL
Each for $1,186.00
Only null left
Add to Cart
 
MS1528S
View Documents
Epredia™
MS1528S
Concentrated 1 mL
Each for $1,866.00
Only null left
Add to Cart
 
MS1528R7
View Documents
Epredia™
MS1528R7
Prediluted 7 mL
Each for $1,530.00
Only null left
Add to Cart
 
MS1528RQ
View Documents
Epredia™
MS1528RQ
Ready-to-Use 12 mL
Each for $1,139.00
Only null left
Add to Cart
 
MS1528R1
View Documents
Epredia™
MS1528R1
Prediluted 1 mL N/A N/A N/A
Description

Description

CD4, a single chain transmembrane glycoprotein, is found on a T cell subset (helper/inducer) representing 45% of peripheral blood lymphocytes. It is also present on 80% of thymocytes and at a lower level on monocytes. It is involved in recognition of antigen presented along with MHC class II by APCs. It serves as receptor for HIV.

Host Species: Mouse

Clone: 4B12

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 59 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CD4 Ab-8
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Stimulated human leukocytes
4B12
Monoclonal
Mouse
T-Cell
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.