Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Antibody (34930), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB312749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312749 Supplier Novus Biologicals Supplier No. FAB379PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone 34930
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD4, Extracellular domain
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.