Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Antibody (GK1.5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP225191SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225191SS 0.025 mg
NBP225191 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225191SS Supplier Novus Biologicals Supplier No. NBP225191SS
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 5 publications

CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), Block/Neutralize.

Specifications

Antigen CD4
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, Neutralization
Classification Monoclonal
Clone GK1.5
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/ 1 million cells. Use reported in scientific literature (PMID 25340039), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay reported in scientific literature (PMID 25340039), In vivo assay reported in scientific literature (PMID 25340039), Flow (Cell Surface) 2.5 ug/ 1 million cells, Block/Neutralize reported in scientific literature (PMID 25340039)
Gene Accession No. NP_000607
Gene Alias CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Gene Symbols CD4
Host Species Rat
Immunogen The mouse cytotoxic T lymphocyte line V4 was used as the immunogen for the CD4 antibody.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 920
Test Specificity This product reacts with Mouse.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.