Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (GK1.5) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 5 publications
$206.50 - $440.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | GK1.5 |
| Concentration | 1.0 mg/ml |
| Dilution | Flow Cytometry 2.5 ug/ 1 million cells. Use reported in scientific literature (PMID 25340039), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay reported in scientific literature (PMID 25340039), In vivo assay reported in scientific literature (PMID 25340039), Flow (Cell Surface) 2.5 ug/ 1 million cells, Block/Neutralize reported in scientific literature (PMID 25340039) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, Neutralization |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP225191SS
![]() |
Novus Biologicals
NBP225191SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP225191
![]() |
Novus Biologicals
NBP225191 |
0.1 mg |
Each for $440.00
|
|
|||||
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), Block/Neutralize.Specifications
| CD4 | |
| 1.0 mg/ml | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, Neutralization | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| NP_000607 | |
| 920 | |
| The mouse cytotoxic T lymphocyte line V4 was used as the immunogen for the CD4 antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| GK1.5 | |
| Flow Cytometry 2.5 ug/ 1 million cells. Use reported in scientific literature (PMID 25340039), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay reported in scientific literature (PMID 25340039), In vivo assay reported in scientific literature (PMID 25340039), Flow (Cell Surface) 2.5 ug/ 1 million cells, Block/Neutralize reported in scientific literature (PMID 25340039) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| CD4 | |
| IgG κ | |
| Protein G purified | |
| This product reacts with Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title