Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Antibody (GK1.5), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB174058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174058 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174058 Supplier Novus Biologicals Supplier No. FAB554J
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Cell depletion, Costimulation of T Cells, CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition of T Cell Function.

Specifications

Antigen CD4
Applications Cell Depletion, Costimulation, CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone GK1.5
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Host Species Rat
Immunogen Mouse CTL clone V4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.