Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Antibody (RPA-T4) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP225199SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225199SS 0.025 mg
NBP225199 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225199SS Supplier Novus Biologicals Supplier No. NBP225199SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 12 publications

CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay, Immunohistochemistry-Paraffin (Negative), CyTOF-ready, Block/Neutralize.

Specifications

Antigen CD4
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), In vitro Assay, Immunohistochemistry (Paraffin), CyTOF, Neutralization
Classification Monoclonal
Clone RPA-T4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin (Negative), CyTOF-ready, Block/Neutralize
Gene Accession No. NP_000607
Gene Alias CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Gene Symbols CD4
Host Species Mouse
Immunogen PHA-stimulated human PBMC.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 920
Test Specificity This product reacts with Human.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.