Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Antibody (YNB46.1.8 (Campath-9H)), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB275900APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB275900APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB275900APC Supplier Novus Biologicals Supplier No. NBP275900APC
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone YNB46.1.8 (Campath-9H)
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Gene Symbols CD4
Host Species Rat
Immunogen Rat T-cell line (NB2) transfected with the human CD4
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 920
Test Specificity Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.