Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human , Alexa Fluor 647, Clone: OKT4, BD Biosciences™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 566681

 View more versions of this product

Catalog No. BDB566681


Only null left
Add to Cart

Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD4
Monoclonal
Alexa Fluor 647
CD4 antigen (p55); T-cell surface antigen T4/Leu-3
Sheep Erythrocyte Rosette-purified Human T Cells
RUO
Human
IgG2b κ
Flow Cytometry
OKT4
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.