Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Mouse anti-Human , Alexa Fluor 647, Clone: OKT4, BD Biosciences™
Mouse Monoclonal Antibody
$128.60 - $279.50
Specifications
Antigen | CD4 |
---|---|
Clone | OKT4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifications
CD4 | |
Flow Cytometry | |
Alexa Fluor 647 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
Primary |
OKT4 | |
Monoclonal | |
Mouse | |
Human | |
CD4 antigen (p55); T-cell surface antigen T4/Leu-3 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title