Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Ligand/TNFSF5 Antibody (hu5c8 (Ruplizumab)), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N275907A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N275907A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N275907A405 Supplier Novus Biologicals Supplier No. NBP275907AF405
Add to Cart
Add to Cart

Human Monoclonal Antibody

CD40 Ligand/TNFSF5 Monoclonal specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CD40 Ligand/TNFSF5
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone hu5c8 (Ruplizumab)
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Gene Symbols CD40LG
Host Species Human
Immunogen Human CD40L
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 959.0
Test Specificity The antibody binds to CD40L with an IC50 of 0.845 ug/ml.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.