Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Ligand/TNFSF5 Antibody (hu5c8 (Ruplizumab)), Alexa Fluor™ 750 - Chimeric, Novus Biologicals™
SDP

Catalog No. NP22678A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22678A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22678A70 Supplier Novus Biologicals Supplier No. NBP252678AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD40 Ligand/TNFSF5 Monoclonal specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CD40 Ligand/TNFSF5
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone hu5c8 (Ruplizumab)
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Gene Symbols CD40LG
Host Species Rabbit
Immunogen Human CD40L
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 959
Test Specificity The antibody binds to CD40L with an IC50 of 0.845 ug/ml.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.