Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Ligand/TNFSF5 Antibody (IDEC-131 (Toralizumab)), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB153147 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153147 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153147 Supplier Novus Biologicals Supplier No. NBP312016IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD40 Ligand/TNFSF5 Monoclonal antibody specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen CD40 Ligand/TNFSF5
Applications Immunofluorescence
Classification Monoclonal
Clone IDEC-131 (Toralizumab)
Conjugate DyLight 755
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Host Species Rabbit
Immunogen The original monoclonal antibody was generated by immunization with a soluble fusion protein of human gp39 (gp39-CD8).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 959
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.