Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40 Rat anti-Human, Clone: FGK45.5, MilliporeSigma™
Rat Monoclonal Antibody
$197.00 - $499.00
Specifications
| Antigen | CD40 |
|---|---|
| Clone | FGK45.5 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF269025
|
MilliporeSigma
MABF269025UL |
25 μL |
Each for $197.00
|
|
|||||
|
MABF2690100
|
MilliporeSigma
MABF2690100UL |
100 μL |
Each for $499.00
|
|
|||||
Description
CD40 Monoclonal antibody specifically detects CD40 in Human samples. It is validated for Flow Cytometry, Affects Function.Specifications
| CD40 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Purified rat monoclonal antibody IgG2a in PBS without azide. | |
| IgG2a κ | |
| Protein G purified |
| FGK45.5 | |
| Flow Cytometry, Functional Assay | |
| Unconjugated | |
| Rat | |
| Human | |
| Recombinant mouse CD40 fusion protein. | |
| Primary | |
| Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title