Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40/TNFRSF5 Antibody (4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB067481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067481 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067481 Supplier Novus Biologicals Supplier No. NBP289639AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen CD40/TNFRSF5
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate Alexa Fluor 350
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD40/TNFRSF5 extracellular domain (NP_001241.1; Met 1-Arg 193).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 958
Test Specificity No cross-reactivity in ELISA with: Human CD27/ TNFRSF7 Mouse CD27/ TNFRSF7 Human CD30/ TNFRSF8 Human DR6/ TNFRSF21 Human CD137/ 4-1BB Human TNFRSF1B/ CD120b Human HVEM/ TNFRSF14 Mouse HVEM/ TNFRSF14 Human RELT/ TNFRSF19L
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.