| Antigen |
CD40/TNFRSF5 |
| Applications |
Flow Cytometry, ELISA, Immunohistochemistry |
| Classification |
Monoclonal |
| Clone |
5D12 |
| Conjugate |
Alexa Fluor 405 |
| Dilution |
Flow Cytometry, ELISA, Immunohistochemistry |
| Formulation |
50mM Sodium Borate |
| Gene Alias |
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 |
| Gene Symbols |
CD40 |
| Host Species |
Human |
| Immunogen |
Anti-CD40/TNFRSF5 mAb 5D12, was generated by immunizing mice with sf9 insect cells expressing recombinant human CD40/TNFRSF5 and selected for the ability to bind EBV-immortalized human B cells. The variable regions from mu5D12 were cloned and used to construct chimeric humanized IgG4 5D12. |
| Purification Method |
Protein A purified |
| Quantity |
0.1 mL |
| Regulatory Status |
RUO |
| Research Discipline |
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
958 |
| Test Specificity |
ch5D12 binds specifically to recombinant and native human CD40/TNFRSF5 ectodomain and has been shown to also bind CD40/TNFRSF5 in cynomolgus and marmoset monkeys. The antibody requires residues within D1 and D1/B2 for binding (Hager et al, 2003) (D1 domain is crucial for CD40L binding). Binding characteristics are very similar between humanized ch5D12 and mu5D12. CD40/TNFRSF5 is a glycoprotein of the TNFR superfamily and is expressed on all mature B cells, dendritic cells, activated monocytes, some endothelial cells and some epithelium including the thymus. CD40/TNFRSF5 binds to CD40L, and this interaction is involved in B-cell activation and proliferation, antigen-presenting cell (APC) activation, initiation of antigen-specific T-cell responses, immunoglobulin production, activation of effector macrophages isotype switching, homotypic adhesion and rescue from apoptosis. |
| Target Species |
Human, Cynomolgus Monkey |
| Content And Storage |
Store at 4°C in the dark. |
| Form |
Purified |
| Isotype |
IgG4 κ |
|
Show More
Show Less
|
|