Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40/TNFRSF5 Antibody (65924), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB173963 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173963 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173963 Supplier Novus Biologicals Supplier No. FAB4401K
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA (Matched Antibody Pair)

Specifications

Antigen CD40/TNFRSF5
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 65924
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD40/TNFRSF5, Val24-Arg193, Accession # P27512
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 958
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.