Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40/TNFRSF5 Antibody (A54 scFv), FITC, Novus Biologicals™
SDP

Catalog No. NB016102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016102 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016102 Supplier Novus Biologicals Supplier No. NBP281101F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD40/TNFRSF5
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone A54 scFv
Conjugate FITC
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Gene Symbols CD40
Host Species Rabbit
Immunogen A54 human anti-CD40/TNFRSF5 scFv was selected from the n-CoDeR phage display library after 4 rounds of selections using biotinylated CD40-Fcgamma fusion protein as bait and subsequent capture of bound phages on streptavidin-caoted magnetic beads.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 958
Test Specificity A54 binds specifically to the D1 domain of CD40/TNFRSF5 (and can only bind to the intact i.e. non-truncated version of CD40/TNFRSF5 and when an AD2 peptide tag has not been added) and can recognize CD40/TNFRSF5 on a B cell line (BJAB). This scFv binds to CD40/TNFRSF5 (immobilized by amine coupling) with a KD within the range of 40nM. CD40/TNFRSF5 is a glycoprotein of the TNFR superfamily and is expressed on all mature B cells, dendritic cells, activated monocytes, some endothelial cells and some epithelium including the thymus. CD40/TNFRSF5 binds to the membrane protein CD154 of the TNF superfamily and this interaction is required for the seconfary immune responses and germinal centre formation.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.