Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40/TNFRSF5 Antibody (G28.5), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB343191 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343191 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343191 Supplier Novus Biologicals Supplier No. NBP252697DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOF

Specifications

Antigen CD40/TNFRSF5
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOF
Classification Monoclonal
Clone G28.5
Conjugate DyLight 594
Dilution Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Host Species Mouse
Immunogen Human erythrocyte-rosette-negative tonsillar lymphocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 958
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.