Learn More
Description
Specifications
Specifications
| Antigen | CD40/TNFRSF5 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI8B8 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 |
| Gene Symbols | CD40 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
