Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD41/Integrin alpha 2b Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501734417 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
Catalog No. Quantity
50-173-4417 150 μL
1 options

Catalog No. 50-173-4417

Supplier: Proteintech Group Inc 245521AP150UL

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD41 (platelet glycoprotein IIb, ITGA2B) is composed of two subunits -120 kDa a, alpha and 23 kDa b, beta- that interact with CD61 in the presence of calcium to form a functional adhesive protein receptor. CD41 is also involved in blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to a variety of proteins including von Willebrand factor, fibrinogen, fibronectin and vitronectin. CD41 is mainly expressed on megakaryocyte-platelet lineage, but generally belongs to the antigens that are expressed during the early stages of hematopoietic differentiation. Diseases associated with CD41 dysfunction include Glansmann Thrombasthenia, and Platelet type-16 bleeding disorder.

Specifications

Antigen CD41/Integrin alpha 2b
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ITGA2B
Gene Accession No. P08514, Q9QUM0
Gene Alias CD41, CD41B, GP2B, GPalpha IIb, GPIIb, GTA, HPA3, Integrin alpha IIb, ITGA2B, ITGAB
Gene Symbols Itga2b
Host Species Rabbit
Immunogen CD41/Integrin alpha 2b Fusion Protein Ag19459
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16399, 3674, 685269
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.