Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42b/GPIb alpha Antibody (486805), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB370888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370888 Supplier Novus Biologicals Supplier No. FAB4067Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen CD42b/GPIb alpha
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 486805
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD42b/GPIb alpha , His17-Leu505, Accession # P07359
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2811
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.