Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42b/GPIb alpha Antibody (AK2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017556 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017556 0.025 mg
NB10063790 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017556 Supplier Novus Biologicals Supplier No. NB100637900.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen CD42b/GPIb alpha
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone AK2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation PBS (pH 7.4) with 0.09% Sodium Azide
Gene Accession No. P07359
Gene Alias Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit
Gene Symbols GP1BA
Host Species Mouse
Immunogen Made to Human CD42b
Molecular Weight of Antigen 69 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2811
Test Specificity Recognises the human CD42b cell surface antigen, also known as platelet glycoprotein GP1B. CD42b is expressed by platelets and megakaryocytes. Clone AK2 has been reported to block the binding of von Willebrand Factor (VWF) to platelets (1).
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.