Learn More
Description
Specifications
Specifications
| Antigen | CD42b/GPIb alpha |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | MM2/174 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin Validated for IHC-P from a verified customer review. |
| Gene Alias | Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit |
| Gene Symbols | GP1BA |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
