Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD42d Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | CD42d |
---|---|
Concentration | 500 μg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119035
|
Invitrogen™
PA5119035 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122111
|
Invitrogen™
PA5122111 |
20 μL |
Each for $178.40
|
|
|||||
Description
CD42d Polyclonal Antibody for Western Blot, IHC (P)
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.Specifications
CD42d | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Gp5 | |
CD42d; Glycoprotein 5; glycoprotein 5 (platelet); glycoprotein 5, platelet; glycoprotein V (platelet); glycoprotein V platelet; GP5; GPV; Platelet glycoprotein 5; platelet glycoprotein V; Platelete glycoprotein 5; Platelets GP V; PLGPV | |
Gp5 | |
Primary | |
Antigen Affinity Chromatography |
500 μg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P40197 | |
2814 | |
Recombinant protein Cluster of Differentiation 42d. The antigen corresponds to amino acid range 17-242 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title