Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD43/Sialophorin Antibody (R2/60), Novus Biologicals™
SDP

Catalog No. NBP1434135 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1434135 0.5 mg
NBP14341301 0.1 mg
NBP14341305 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP1434135 Supplier Novus Biologicals Supplier No. NBP1434130.5MG

Rat Monoclonal Antibody

CD43/Sialophorin Monoclonal specifically detects CD43/Sialophorin in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen CD43/Sialophorin
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone R2/60
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500
Formulation Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Gene Alias CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43)
Gene Symbols SPN
Host Species Rat
Immunogen The immunogen for this antibody was CD43.
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6693
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.