Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD43/Sialophorin Antibody (R2/60), Novus Biologicals™
SDP

Rat Monoclonal Antibody

$305.00 - $611.00

Specifications

Antigen CD43/Sialophorin
Clone R2/60
Concentration 0.5 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500
Applications Western Blot, Flow Cytometry, Immunoprecipitation
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14341305
SDP
View Documents
Novus Biologicals
NBP1434130.05MG
0.05 mg
Each for $305.00
Only null left
Add to Cart
 
NBP14341301
SDP
View Documents
Novus Biologicals
NBP1434130.1MG
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
NBP1434135
SDP
View Documents
Novus Biologicals
NBP1434130.5MG
0.5 mg
Each for $611.00
Only null left
Add to Cart
 
Description

Description

CD43/Sialophorin Monoclonal specifically detects CD43/Sialophorin in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.
Specifications

Specifications

CD43/Sialophorin
0.5 mg/mL
Western Blot, Flow Cytometry, Immunoprecipitation
Unconjugated
RUO
Mouse
CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43)
SPN
IgM
Affinity Purified
R2/60
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500
Monoclonal
Rat
B Cell Development and Differentiation Markers, Immunology
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
6693
The immunogen for this antibody was CD43.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.