Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD43/Sialophorin Antibody (W3/13), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB174767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174767 Supplier Novus Biologicals Supplier No. FAB10388Y
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD43/Sialophorin Monoclonal antibody specifically detects CD43/Sialophorin in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen CD43/Sialophorin
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone W3/13
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43)
Host Species Mouse
Immunogen Rat thymocyte membrane glycoproteins, Accession # P13838
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6693
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.