Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Antibody (065), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB037290 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037290 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037290 Supplier Novus Biologicals Supplier No. NBP290077FR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CD44
Applications Western Blot, ELISA
Classification Monoclonal
Clone 65
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD44 (P16070-1; Met 1-Pro 220).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cell Biology, Core ESC Like Genes, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 960
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.