Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Antibody (8E2F3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP147386SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147386SS 0.025 mL
NBP147386 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP147386SS Supplier Novus Biologicals Supplier No. NBP147386SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 14 publications

CD44 Monoclonal specifically detects CD44 in Human, Mouse, Rabbit (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen CD44
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8E2F3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:1000, Flow (Intracellular)
Gene Accession No. P16070
Gene Alias CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
Gene Symbols CD44
Host Species Mouse
Immunogen Purified recombinant fragment of human CD44 (628-699) expressed in E. coli. [Uniprot: P16070]
Molecular Weight of Antigen 82 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 960
Test Specificity Not reactive to rabbit per customer review. Immunogen's sequence homology with other species: Bovine (89%), Rat (87%), Golden hamster (87%) and Chinese hamster (80%).
Target Species Human, Mouse, Rabbit (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.