Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP22253050UG
Description
CD44 Monoclonal specifically detects CD44 in Human, Mouse, Bovine, Goat, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready, Block/Neutralize.Specifications
CD44 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:50, CyTOF-ready, Neutralization | |
CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This antibody reacts with Ovine. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF, Neutralization | |
Hermes-1 | |
Unconjugated | |
P16070 | |
CD44 | |
80 - 95 kDa lymphocyte surface glycoprotein H-CAM (CD44). | |
50 ug | |
Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
960 | |
Human, Mouse, Bovine, Goat, Rabbit, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction