Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD44 Antibody (Hermes-1) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 4 publications
$196.10 - $512.00
Specifications
Antigen | CD44 |
---|---|
Clone | Hermes-1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:50, CyTOF-ready, Neutralization |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF, Neutralization |
Description
CD44 Monoclonal specifically detects CD44 in Human, Mouse, Bovine, Goat, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready, Block/Neutralize.Specifications
CD44 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF, Neutralization | |
Unconjugated | |
Rat | |
Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
P16070 | |
960 | |
80 - 95 kDa lymphocyte surface glycoprotein H-CAM (CD44). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Hermes-1 | |
Western Blot 1:1 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 1:10 - 1:50, CyTOF-ready, Neutralization | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Bovine, Goat, Rabbit, Sheep | |
CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
CD44 | |
IgG2a | |
Protein G purified | |
This antibody reacts with Ovine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title