Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Antibody (VFF-17), DyLight 755 - variant 7/8, Novus Biologicals™
SDP

Catalog No. NB10065535I Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065535I 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065535I Supplier Novus Biologicals Supplier No. NB10065535IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen CD44
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VFF-17
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. P16070
Gene Alias CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
Gene Symbols CD44
Host Species Mouse
Immunogen Made to Human CD44v7/8
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 960
Test Specificity Specifically recognizes an epitope encoded by exons v7-v8 on the variant portion of human CD44.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.