Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Antibody (30-F11), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB305198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305198 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305198 Supplier Novus Biologicals Supplier No. FAB114MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry, CyTOF-reported

Specifications

Antigen CD45
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cytotoxicity Assay, Immunocytochemistry
Classification Monoclonal
Clone 30-F11
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen
Host Species Rat
Immunogen Mouse thymus and spleen cells
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.