Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Antibody (F10-89-4), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB004352 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004352 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004352 Supplier Novus Biologicals Supplier No. NBP128509JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45 Monoclonal specifically detects CD45 in Human, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.

Specifications

Antigen CD45
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone F10-89-4
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Mouse
Immunogen Purified T cells from human lymph nodes
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Test Specificity Human CD45
Target Species Human, Equine
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.