Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Antibody (LT40), Novus Biologicals™
SDP

Catalog No. NBP128295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128295 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128295 Supplier Novus Biologicals Supplier No. NBP128295

Mouse Monoclonal Antibody

CD45 Monoclonal specifically detects CD45 in Chicken samples. It is validated for Flow Cytometry.

Specifications

Antigen CD45
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone LT40
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Mouse
Immunogen Chicken bursal lymphocytes
Purification Method IgM purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Test Specificity Chicken CD45 (Mr 190 to 215-kDa).
Target Species Chicken
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.